missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFR alpha-2 Monoclonal Antibody (71515)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561631
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
Specifications
| GFR alpha-2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O00451 | |
| GFRA2 | |
| Mouse myeloma cell line NS0-derived recombinant human GFR alpha -2/GDNF R alpha -2, Ser22-Ser441. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| 71515 | |
| Unconjugated | |
| GFRA2 | |
| GDNF family receptor alpha 2; GDNF family receptor alpha-2; GDNF re; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; Gdnfrb; GDNFR-beta; Gfra2; GFR-alpha 2; GFR-alpha-2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; HpV18gp1; neurturin receptor alpha; NRTNR-alpha; NTNRA; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; RETL2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; TRNR2; tyrosine kinase receptor ligand 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction