Learn More
GH2 Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811356S
Description
GH2 Monoclonal antibody specifically detects GH2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| GH2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13105 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7709 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B11 | |
| Unconjugated | |
| ZBTB17 | |
| GH2; GHB2; GHL; GHV; GH-V; Growth hormone 2; growth hormone B2; growth hormone variant; hGH-V; placental-specific growth hormone; Placenta-specific growth hormone | |
| Human recombinant protein fragment corresponding to amino acids 112-457 of human ZBTB17 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.