missing translation for 'onlineSavingsMsg'
Learn More

Ginsenoside C-K, TRC

Product Code. 30240742
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240742 5 mg 5mg
30240650 50 mg 50mg
30240628 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240742 Supplier Toronto Research Chemicals Supplier No. TRCG4102805MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39262-14-1
Molecular Formula C36 H62 O8
Molecular Weight (g/mol) 622.87
Synonym β-D-Glucopyranoside, (3β,12β)-3,12-dihydroxydammar-24-en-20-yl, (3β,12β)-3,12-Dihydroxydammar-24-en-20-yl β-D-glucopyranoside, 20(S)-Protopanaxadiol 20-O-D-glucopyranoside, 20(S)-Protopanaxadiol 20-O-β-D-glucopyranoside, 20-O-β-D-Glucopyranosyl-20(S)-protopanaxadiol, 20-O-β-Glucopyranosyl-20(S)-protopanaxadiol, 3-O-Deglucosylginsenoside F2, Compound k, Ginsenoside C-K, Ginsenoside IH901, Ginsenoside K, Ginsenoside M1, Ginsenoside compound K, Ginsenoside metabolite M1, IH 901, Protopanaxadiol 20-O-glucoside
IUPAC Name (2S,3R,4S,5S,6R)-2-[(2S)-2-[(3S,5R,8R,9R,10R,12R,13R,14R,17S)-3,12-dihydroxy-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-methylhept-5-en-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CC(=CCC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C36H62O8/c1-20(2)10-9-14-36(8,44-31-30(42)29(41)28(40)23(19-37)43-31)21-11-16-35(7)27(21)22(38)18-25-33(5)15-13-26(39)32(3,4)24(33)12-17-34(25,35)6/h10,21-31,37-42H,9,11-19H2,1-8H3/t21-,22+,23+,24-,25+,26-,27-,28+,29-,30+,31-,33-,34+,35+,36-/m0/s1
Formula Weight 622.4445
Chemical Name or Material Ginsenoside K
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.