missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIP Antibody (4), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21617DL594
This item is not returnable.
View return policy
Description
GIP Monoclonal antibody specifically detects GIP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GIP | |
| Monoclonal | |
| DyLight 594 | |
| gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide | |
| Synthetic human gastric inhibitory polypeptide | |
| 0.1 mL | |
| Cell Biology, GPCR, Signal Transduction | |
| 2695 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction