missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GJC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55376-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| GJC2 | |
| Polyclonal | |
| Unconjugated | |
| GJC2 | |
| Connexin 46.6, Connexin 47, CX46.6, Cx47, Gap junction alpha 12 protein, Gap junction gamma 2 protein, GJA12, GJC2, HLD2, PMLDAR, SPG44 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 118454, 57165 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5T442, Q8BQU6 | |
| Gjc2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction