missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GK5 Monoclonal Antibody (2C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523216
This item is not returnable.
View return policy
Description
Peptide Sequence: MSGLLTDPEQ RAQEPRYPGF VLGLDVGSSV IRCHVYDRAA RVCGSSVQKV ENLYPQIGWV EIDPDVLWIQ FVAVIKEAVK AAGIQMNQI.
GK5 (Glycerol Kinase 5) is a Protein Coding gene. Diseases associated with GK5 include Type 1 Diabetes Mellitus 7 and Type 1 Diabetes Mellitus 13. Among its related pathways are glycerol degradation and triacylglycerol biosynthesis. Gene Ontology (GO) annotations related to this gene include phosphotransferase activity, alcohol group as acceptor and glycerol kinase activity. An important paralog of this gene is GK. [GeneCards].
Specifications
| GK5 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q6ZS86 | |
| GK5 | |
| GK5 (NP_689989, 1 a.a. ~ 89 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2C11 | |
| Unconjugated | |
| GK5 | |
| ATP:glycerol 3-phosphotransferase 5; AV095337; C330018K18Rik; G630067D24Rik; GK 5; Gk5; glycerokinase 5; glycerol kinase 5 (putative); putative glycerol kinase 5; RGD1311223 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 256356 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction