Learn More
RETSCH Glass Beads for Mills
Product Code. 11756175 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
0.10 to 0.25 mm
0.25 to 0.50 mm
0.50 to 0.75 mm
0.75 to 1 mm
1 to 1.5 mm
Unit Size:
500g
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11756175 0.75 to 1 mm 500g
11726175 0.10 to 0.25 mm Each
11736175 0.25 to 0.50 mm Each
10782701 1 to 1.5 mm 500g
11746175 0.50 to 0.75 mm 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11756175 Supplier RETSCH Supplier No. 22.222.0004
  • ONLINE EXCLUSIVE PRICE
    61.15€
    59.68€ / 500g
    Save 1.47€
    2% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Retsch MM 200 Mixer Mills.
  • Ideal for disruption of biological cells
Specifications
Type Beads
Material Glass
Diameter (Metric) 0.75 to 1 mm
For Use With (Equipment) Mixer Mill MM 200
Quantity 500 g