Learn More
Marienfeld Superior™ Glass Capillary Tubes for the Determination of Melting Point
Product Code. 10764034
Change view
Click to view available options
Length (Metric) Tube:
70 mm
75 mm
80 mm
Diameter (Metric) Inner:
0.42 mm
0.9 mm
1 mm
1.2 mm
1.5 mm
Product Type:
Capillary Tubes (Both Ends Open)
Capillaty Tubes (One End Closed)
Unit Size:
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tube Diameter (Metric) Inner Product Type Unit Size
10764034 80 mm 1.2 mm Capillaty Tubes (One End Closed) Pack of 1000
10640783 80 mm 1.2 mm Capillary Tubes (Both Ends Open) Pack of 1000
10185413 80 mm 1 mm Capillaty Tubes (One End Closed) Pack of 1000
10222413 80 mm 0.42 mm Capillary Tubes (Both Ends Open) Pack of 1000
10501363 70 mm 0.9 mm Capillary Tubes (Both Ends Open) Pack of 1000
16341238 75 mm 1.5 mm Capillaty Tubes (One End Closed) Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10764034 Supplier Marienfeld Superior™ Supplier No. 2940207
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use Marienfeld Superior™ Glass Capillary Tubes for storage of small amounts of liquids and to determine melting points during laboratory experimentation.
  • With rectangular cut edges
  • For single use
  • 100 capillaries in a dispenser vial, 10 vials in a box
Specifications
Material Glass
Product Type Capillaty Tubes (One End Closed)
Diameter (Metric) Inner 1.2 mm
For Use With (Application) For Determination Of Melting Point
Length (Metric) Tube 80 mm
Quantity 1000