Learn More
Alla France™ Glass Hydrometer
Product Code. 11793183
Change view
Click to view available options
Specific Gravity Range:
0.800 to 0.900 g/mL
1.000 to 1.100 g/mL
1.100 to 1.200 g/mL
1.200 to 1.300 g/mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Specific Gravity Range Unit Size
11793183 1.000 to 1.100 g/mL Each
11703193 1.100 to 1.200 g/mL Each
11773183 0.800 to 0.900 g/mL Each
11713193 1.200 to 1.300 g/mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11793183 Supplier Alla France™ Supplier No. 3010FG100/20qp
  • ONLINE EXCLUSIVE PRICE
    38.97€
    14.80€ / Pack of 1
    Save 24.17€
    62% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use these hydrometers for a variety of applications. Alla France™ Glass Hydrometers feature a benchmark engraved on the stem to show any displacement of the scale.
  • General Purpose Hydrometers
  • 100 g/mL series
  • Density scale (g/mL)
  • Reading below meniscus
  • Without Certificate
Specifications
Quantity 1Each
Type Density Hydrometer
Accuracy ±0.002g/mL
Increments 0.001 g/mL
Length (Metric) 300 mm
For Use With (Application) General use
Specific Gravity Range 1.000 to 1.100 g/mL