missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade MGA

Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.

Brand:  Sartorius Stedim Biotech™ FT-2-1101-620620

Additional Details : Weight : 1.99800kg

 View more versions of this product

Product Code. 15992720

  • 1350.00€ / Pack of 100
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Binder-free
  • Suitable for the following:
    • Air/gas filtration
    • Liquid filtration
    • Clarification of buffer and reagent solutions
    • Analytical sample preparation
    • Particle analysis
  • Non-sterile, bulk packed
Specifications

Specifications

55 g/m2
Sheet
1.6 μm
510 mL/min.
Hydrophilic
0.25 mm
White
Binder-free Glass Microfiber
Non-sterile
MGA
100/Pk.
620 x 620 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions