missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade MGA
Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.
119.00€ - 1350.00€
Specifications
Basis Weight | 55 g/m2 |
---|---|
Color | White |
Format | Sheet |
Material | Binder-free Glass Microfiber |
Particle Retention | 1.6 μm |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15972720
|
Sartorius Stedim Biotech™
FT-2-1101-103257 |
103 x 257 mm |
119.00€
Pack of 100 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15982720
|
Sartorius Stedim Biotech™
FT-2-1101-203254 |
203 x 254 mm |
308.00€
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15992720
|
Sartorius Stedim Biotech™
FT-2-1101-620620 |
620 x 620 mm |
1350.00€
Pack of 100 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Binder-free
- Suitable for the following:
- Air/gas filtration
- Liquid filtration
- Clarification of buffer and reagent solutions
- Analytical sample preparation
- Particle analysis
- Non-sterile, bulk packed
Specifications
55 g/m2 | |
Sheet | |
1.6 μm | |
510 mL/min. | |
Hydrophilic | |
0.25 mm |
White | |
Binder-free Glass Microfiber | |
Non-sterile | |
MGA | |
100/Pk. |