Learn More
Cole-Parmer™ Glass Mortar
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
100mL
Specifications
Specifications
| Capacity (Metric) | 2 mL |
| Product Type | Glass Mortar |
| Material | Glass |
| Type | Glass Mortar |