Learn More
Macherey-Nagel™ Glass Plate Nano-Adamant UV254
Product Code. 10661553 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10661553 10 x 20 cm Pack of 50
10466912 10 x 10 cm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10661553 Supplier Macherey-Nagel™ Supplier No. 821120
 Restricted Item
Estimated Shipment: 12-08-2026
Specifications
Particle Size 2 to 10 μm
Pore Size 60 Å
Quantity 50 Pk.
Type Glass Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 10 x 20 cm