Learn More
SHP Steriltechnik AG Glass Test Programme
Product Code. 31908845
Change view
Click to view available options
For Use With (Equipment):
Autoclaves
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31908845 Autoclaves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31908845 Supplier SHP Steriltechnik AG Supplier No. 20.0000.000.140
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Special programme is designed for the gentle sterilisation of glassware in an autoclave. It prevents thermal stress and breakage whilst ensuring reliable sterility.
  • Sterilisation of glass containers, bottles and test tubes
  • Gentle heating and cooling to prevent breakage
  • Uniform steam penetration for reliable sterility
  • Reproducible results for laboratory and research applications
  • Simple operation via autoclave control panel
Specifications
For Use With (Equipment) Autoclaves
Product Type Software