Learn More
Rogo Sampaic™ Class B Glass Volumetric Pipettes
Product Code. 11722684
Click to view available options
Volume (Metric):
2 mL
10 mL
20 mL
25 mL
50 mL
Color:
Black
Orange
Red
Yellow
Unit Size:
Each
Pack of 2
Product Code. 11722684
Brand: Rogo Sampaic™ DGG007
Only null left
Add to basket
Edge
Add to basket
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Rogo Sampaic™ Class B Glass Volumetric Pipettes are manufactured from clear soda lime glass.
  • With vitrified amber graduations
  • With safety bulb
Specifications
Calibrated Yes
Calibration 0.1mL
Class B
Graduated Yes
Graduation Color Brown
Graduations 0.1 mL
Color Orange
Volume (Metric) 50 mL
Length (Metric) 700 mm