Learn More
Karl Hecht™ Assistent™ Glass Wool
Product Code. 10155231
Change view
Click to view available options
Weight (Metric):
1kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
10155231 1kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10155231 Supplier Karl Hecht™ Supplier No. 41408003
  • ONLINE EXCLUSIVE PRICE
    49.07€
    32.83€ / 1kg
    Save 16.24€
    33% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Captures even extremely small particles during filtration. Karl Hecht™ Assistent™ Glass Wool is soft, strong and superfine.
  • Suitable for filtration
  • White in color
  • Ideal for general lab and industrial applications
Specifications
Weight (Metric) 1kg