Learn More
Smeg™ Glassware GW4090C Washer with Drying System and Steam Condenser
Product Code. 11952699
Change view
Click to view available options
:
Glassware GW4090C Washer with Drying System and Steam Condenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11952699 Glassware GW4090C Washer with Drying System and Steam Condenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11952699 Supplier Smeg™ Supplier No. GW4090C
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Used for antibacterical thermal disinfection. Smeg Instruments Laboratory Glassware GW4090C Washer with Drying System and Steam Condenser has electronic door lock system for user safety.
  • Washing up to 95°C
  • 3 Microprocessor programmable electronic control
  • Serial port RS232 for connection to external printer or PC
  • 128 x 64 pixel graphics display for different languages messages
  • High efficency steam condenser with cold water injection
  • Active thermodynamic drying
  • High efficency steam condenser Ecoslim, with cold water injection
  • Peristaltic pump for liquid detergent, liquid neutralizer
  • Installing two additional peristaltic pump possible
  • Washing chamber and internal door is made of AISI 316L stainless steel
  • External panels are made of AISI 304 stainless steel
  • Two independent washing levels through telescopic guides
  • Contains water softener with salt sensor
  • Cold water, hot water and demineralized water connections
  • Contains no trolleys
  • Programs: 9 default, 6 free
  • External dimensions: L x D x H: 900 x 640 x 850mm
  • Internal working dimensions LxDxH: 525 x 490 x 570mm
Specifications
Product Type Glassware Washer with Drying System and Steam Condenser
Model GW4090C
Electrical Requirements 3/N/PE 7kW max
Voltage 400 V
Frequency 50 Hz