missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glassy carbon rod, 7mm (0.28 in.) dia, type 2, Thermo Scientific Chemicals
Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.
355.00€ - 939.00€
Chemical Identifiers
Chemical Name or Material | Glassy carbon rod |
---|---|
CAS | 7440-44-0 |
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374307
|
Thermo Scientific Alfa Aesar
037999.DJ |
50 mm |
355.00€
50mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11384307
|
Thermo Scientific Alfa Aesar
037999.DM |
100 mm |
640.00€
100mm |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15411477
|
Thermo Scientific Alfa Aesar
037999.DR |
200 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used electrical conductor ion-selective membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
Glassy carbon rod | |
Tube | |
Insoluble in water. |
7440-44-0 | |
MFCD00133992 |
Specifications
Glassy carbon rod | |
Tube | |
50 mm |
7440-44-0 | |
MFCD00133992 | |
Insoluble in water. |
RUO – Research Use Only