missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon splinter powder, 0.4 to 12μm, type 2, Thermo Scientific Chemicals

Product Code. 11334327
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334327 50 g 50g
11344327 10 g 10g
11324327 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334327 Supplier Thermo Scientific Alfa Aesar Supplier No. 038007.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as such in purifying metals, in electrodes, electrical devices & steel.

Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Packaging Glass bottle
Quantity 50 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon splinter powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.