missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-1 Antibody (CL12192) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21211G
This item is not returnable.
View return policy
Description
GLI-1 Monoclonal antibody specifically detects GLI-1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| GLI-1 | |
| Monoclonal | |
| DyLight 488 | |
| GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| This antibody was developed using a synthetic peptide derived from P08151, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 2735 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12192 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction