missing translation for 'onlineSavingsMsg'
Learn More

Glibenclamide EP Impurity C (Glipizide EP Impurity I), Mikromol™

Product Code. 30458168
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458168 4 x 25 mg 100mg
17912246 100 mg 100mg
17941646 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458168 Supplier Mikromol Supplier No. MM0942.0900254AMP
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Glibenclamide EP Impurity C (Glipizide EP Impurity I), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
CAS 10079-35-3
Analyte or Component Names N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
Shipping Condition Room Temperature
Molecular Formula C22 H34 N4 O4 S
Certifications/Compliance ISO 9001
Synonym Glibenclamide Imp. C (EP), Glipizide Imp. I (EP), N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide, 1-[[4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea, Glibenclamide Imp. C (Pharmeuropa), Glibenclamide Impurity C, Glipizide Impurity I
InChI Formula InChI=1S/C22H34N4O4S/c27-21(24-18-7-3-1-4-8-18)23-16-15-17-11-13-20(14-12-17)31(29,30)26-22(28)25-19-9-5-2-6-10-19/h11-14,18-19H,1-10,15-16H2,(H2,23,24,27)(H2,25,26,28)
Solution Type Neat
SMILES O=C(NCCc1ccc(cc1)S(=O)(=O)NC(=O)NC2CCCCC2)NC3CCCCC3
IUPAC Name 1-cyclohexyl-3-[4-[2-(cyclohexylcarbamoylamino)ethyl]phenyl]sulfonylurea
Molecular Weight (g/mol) 450.59
Quantity 4 x 25 mg
Formula Weight 450.2301 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.