missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SKIV2L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102808
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MTR4.
SKIV2L2 may be involved in pre-mRNA splicing.
Specifications
| SKIV2L2 | |
| Polyclonal | |
| Unconjugated | |
| MTREX | |
| 2610528A15Rik; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Dob1; Exosome RNA helicase MTR4; exosome RNA helicase MTR4; superkiller viralicidic activity 2-like 2; fSAP118; functional spliceosome-associated protein 118; KIAA0052; mKIAA0052; Mtr4; Mtr4 exosome RNA helicase; MTREX; RGD1305984; Ski2 like RNA helicase 2; SKIV2L2; Superkiller viralicidic activity 2-like 2; superkiller viralicidic activity 2-like 2 (S. cerevisiae); superkiller viralicidic activity 2-like 2-like; TRAMP-like complex helicase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23517, 365668, 72198 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P42285, Q9CZU3 | |
| MTREX | |
| A synthesized peptide derived from human SKIV2L2(Accession P42285), corresponding to amino acid residues S758-P808. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction