missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP1 Antibody (2622D) [Biotin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB124901B
This item is not returnable.
View return policy
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human samples. It is validated for ELISA
Specifications
| GLP1 | |
| Monoclonal | |
| Biotin | |
| glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
| Human GLP-1 synthetic peptide, Accession # P01275 | |
| 0.1 mL | |
| Cancer, Signal Transduction, Stem Cell Markers | |
| 2641 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 2622D | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction