missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLRA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13145-1-AP-20UL
This item is not returnable.
View return policy
Description
The GLRA3 gene encodes the alpha-3 subunit of the neuronal glycine receptor, a ligand-gated chloride channel composed of ligand-binding alpha and structural beta polypeptides.Specifications
| GLRA3 | |
| Polyclonal | |
| Unconjugated | |
| GLRA3 | |
| GLRA3, glycine receptor, alpha 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110304, 114516, 8001 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75311, P24524, Q91XP5 | |
| Glra3 | |
| GLRA3 Fusion Protein Ag3702 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction