missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLSA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598350
Additional Details : Weight : 0.01000kg
Description
Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.Specifications
GLSA1 | |
Polyclonal | |
Unconjugated | |
6330442B14, AAD20, AI314027, B230365M23Rik, DKFZp686O15119, FLJ10358, GA, GAC, GAM, GLS, GLS1, Glut, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, Glutaminase kidney isoform, mitochondrial 65 kDa chain, Glutaminase kidney isoform, mitochondrial 68 kDa chain, glutaminase, phosphate-activated, KGA, K-glutaminase, KIAA0838, L-glutamine amidohydrolase, PAG, phosphate-activated glutaminase, RATGLUT | |
Recombinant Escherichia coli Glutaminase 1 protein (1-310AA) | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |