missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1 Antibody (S355-1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22399-0.025mg
This item is not returnable.
View return policy
Description
GluR1 Monoclonal specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GluR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2890 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S355-1 | |
| Unconjugated | |
| P19490 | |
| GRIA1 | |
| Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1 | |
| 0.025 mg | |
| Primary | |
| Detects approx. 100kDa. Does not cross-react with GluR2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction