missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1, p Ser845 Antibody (RM296), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61472
This item is not returnable.
View return policy
Description
GluR1 Monoclonal antibody specifically detects GluR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GluR1 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM296 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
| AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
| A phospho-peptide corresponding to Human GluR1 [p Ser845] | |
| 100 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
| 2890 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction