missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut2 Antibody (199017), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1414B
This item is not returnable.
View return policy
Description
Glut2 Monoclonal antibody specifically detects Glut2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Glut2 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 199017 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| NS0 mouse myeloma cell line transfected with human Glut2, Met1-Val524, Accession # P11168 | |
| 0.2 mL | |
| Lipid and Metabolism, Stem Cell Signaling Pathway | |
| 6514 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction