missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597263
This item is not returnable.
View return policy
Description
Positive Samples: SW480, THP-1, Mouse liver, Mouse kidney, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: YVINSTDELP TISYSMN.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
Specifications
| GLUT2 | |
| Polyclonal | |
| Unconjugated | |
| Slc2a2 | |
| AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20526, 25351, 6514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P11168, P12336, P14246 | |
| Slc2a2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction