Learn More
Invitrogen™ Talacotuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558479
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Talacotuzumab, also known as JNJ-56022473 (and formerly CSL-362) is a humanized IgG1 monoclonal antibody designed to bind to and neutralize CD123 (Interleukin-3 receptor alpha chain or IL3RA) with potential antineoplastic activity.
Specifications
| Talacotuzumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CSL362; CSL-362-AML; JNJ-56022473 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |