missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Hormone Antibody (GH-45), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-364AF532
This item is not returnable.
View return policy
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Growth Hormone | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GH1 | |
| Human growth hormone | |
| Primary | |
| The antibody GH-45 reacts with human growth hormone (hGH), a polypeptide hormone synthesized by acidophilic or somatotropic cells of the anterior pituitary gland. The GH-45 antibody reacts with affinity constant 3.8 x 1010 l/mol; it does not bind human prolactin or any other pituitary hormones. | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GH-45 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
| Mouse | |
| 0.1 mL | |
| 2688 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction