missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaminase Monoclonal Antibody (9G6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA549191
Additional Details : Weight : 0.01000kg
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.Specifications
Glutaminase | |
Monoclonal | |
500 μg/mL | |
PBS with 4MG trehalose and no preservative | |
D3Z7P3, O94925, P13264 | |
GLS | |
E.coli-derived human Glutaminase/GLS recombinant protein (Position: K396-N654). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
9G6 | |
Unconjugated | |
GLS | |
6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
14660, 24398, 2744 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2b |