missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaminase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5115296
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total Glutaminase.
Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.Specifications
Glutaminase | |
Polyclonal | |
Unconjugated | |
GLS | |
6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT | |
Rabbit | |
Affinity chromatography | |
RUO | |
14660, 24398, 2744 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
D3Z7P3, O94925, P13264 | |
GLS | |
A synthesized peptide derived from human GLS(Accession O94925), corresponding to amino acid residues V592-Y642. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |