missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaminase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535365
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.Specifications
Glutaminase | |
Polyclonal | |
Unconjugated | |
GLS | |
6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14660, 24398, 2744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
D3Z7P3, O94925, P13264 | |
GLS | |
KLH conjugated synthetic peptide between 516-545 amino acids from the C-terminal region of human GLS | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |