missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reductase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99476-50ul
This item is not returnable.
View return policy
Description
Glutathione Reductase Polyclonal antibody specifically detects Glutathione Reductase in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Glutathione Reductase | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| EC 1.8.1, EC 1.8.1.7, GLUR, glutathione reductase, glutathione reductase, mitochondrial, GR, GRase, GRD1, MGC78522 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Glutathione Reductase. | |
| 50 μg | |
| Cancer | |
| 2936 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction