Learn More
Glycerol Kinase Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NB120-20600-0.1MG
Product Code. 18217763
  • 725.00€ / 0.10mg
Description
Glycerol Kinase Polyclonal specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glycerol Kinase
Polyclonal
Western Blot 1:500 - 1:2000, ELISA 1:4000 - 1:20000
ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase
Goat
Delipidation and Defibrination
RUO
2710
Bacteria, Non-species specific
Purified
Western Blot, ELISA
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
GK
Glycerol Kinase [Cellulomonas species]
0.1 mg
Primary
This is specific for purified and partially purified Glycerol Kinase (Cellulomonas species).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.