missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl Oleate (1.0 mg/mL in Acetonitrile), TRC
Specifications
Specifications
| CAS | 5431-33-4 |
| Molecular Formula | C21 H38 O3 |
| Quantity | 10 x 1 mL |
| Synonym | Glycidyl Oleate, Oleic acid-glycidyl ester, 9-Octadecenoic acid (9Z)-, 2-oxiranylmethyl ester, 9-Octadecenoic acid (9Z)-, oxiranylmethyl ester (9CI), 9-Octadecenoic acid (Z)-, oxiranylmethyl ester, Oleic acid, 2,3-epoxypropyl ester (6CI,7CI,8CI), 1-Propanol, 2,3-epoxy-, oleate (8CI), Glycidyl oleate, NSC 13542, Oleic acid glycidyl ester |
| InChI Formula | InChI=1S/C21H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h9-10,20H,2-8,11-19H2,1H3/b10-9- |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC1CO1 |
| IUPAC Name | oxiran-2-ylmethyl (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 338.52 |
| Formula Weight | 338.2821 |
| Concentration or Composition (by Analyte or Components) | Glycidyl Oleate 1 mg/mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?