missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Oleate, TRC

Product Code. 17984135
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984135 100 mg 100mg
17943535 25 mg 25mg
17952285 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984135 Supplier Toronto Research Chemicals Supplier No. TRCG615930100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5431-33-4
Molecular Formula C21 H38 O3
Molecular Weight (g/mol) 338.52
Synonym Glycidyl Oleate, Oleic acid-glycidyl ester, 9-Octadecenoic acid (9Z)-, 2-oxiranylmethyl ester, 9-Octadecenoic acid (9Z)-, oxiranylmethyl ester (9CI), 9-Octadecenoic acid (Z)-, oxiranylmethyl ester, Oleic acid, 2,3-epoxypropyl ester (6CI,7CI,8CI), 1-Propanol, 2,3-epoxy-, oleate (8CI), Glycidyl oleate, NSC 13542, Oleic acid glycidyl ester
IUPAC Name oxiran-2-ylmethyl (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC1CO1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h9-10,20H,2-8,11-19H2,1H3/b10-9-
Formula Weight 338.2821
Percent Purity >95%
Chemical Name or Material Glycidyl Oleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.