missing translation for 'onlineSavingsMsg'
Learn More

Glycine Ethyl Ester Hydrochloride, TRC

Product Code. 30135784
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135784 250 g 250g
30137897 100 g 100g
30003502 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135784 Supplier Toronto Research Chemicals Supplier No. TRCG625225250G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-33-6
Molecular Formula C4 H9 N O2 . Cl H
Molecular Weight (g/mol) 139.58
Synonym Glycine, ethyl ester, hydrochloride (1:1), Glycine, ethyl ester, hydrochloride (6CI,8CI,9CI), (2-Ethoxy-2-oxoethyl)ammonium chloride, Aminoacetic acid ethyl ester hydrochloride, Ethyl 2-aminoacetate hydrochloride, Ethyl aminoacetate hydrochloride, Ethyl glycinate hydrochloride, Glycine ethyl ester hydrochloride, Glycine ethyl ester hydrochloride salt, Glycine ethyl ester monohydrochloride, Glycine ethyl ether hydrochloride, Glycine hydrochloride ethyl ester, L-Glycine ether ester hydrochloride
IUPAC Name ethyl 2-aminoacetate;hydrochloride
SMILES Cl.CCOC(=O)CN

Specifications

InChI Formula InChI=1S/C4H9NO2.ClH/c1-2-7-4(6)3-5;/h2-3,5H2,1H3;1H
Quantity 250 g
Formula Weight 139.04
Shipping Condition Room Temperature
Chemical Name or Material Glycine Ethyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.