missing translation for 'onlineSavingsMsg'
Learn More

Glycochenodeoxycholic Acid Sodium, 95%, TRC

Product Code. 30408064
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408064 100 mg 100mg
30403308 25 mg 25mg
30390722 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408064 Supplier Toronto Research Chemicals Supplier No. TRCG641255100MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycochenodeoxycholic Acid Sodium Salt
CAS 16564-43-5
Purity Grade Notes HPLC
Molecular Formula C26 H42 N O5 . Na
Quantity 100 mg
Synonym N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium, N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Monosodium, NSC 681056, Sodium Chenodeoxycholylglycine, Sodium Glycochenodeoxycholate, Sodium Glycochenodesoxycholate
InChI Formula InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;/m1./s1
SMILES [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
Molecular Weight (g/mol) 471.61
Formula Weight 471.2961
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.