missing translation for 'onlineSavingsMsg'
Learn More

Glycochenodeoxycholic Acid Sodium Salt (Ultra pure, >97%), TRC

Product Code. 30240646
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240646 1 g 1g
30240785 50 mg 50mg
30240669 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240646 Supplier Toronto Research Chemicals Supplier No. TRCG6412601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 16564-43-5
Molecular Formula C26 H42 N O5 . Na
Molecular Weight (g/mol) 471.61
Synonym N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium Salt, N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Monosodium Salt, NSC 681056, Sodium Chenodeoxycholylglycine, Sodium Glycochenodeoxycholate, Sodium Glycochenodesoxycholate;
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
SMILES [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifica

Quantity 1 g
InChI Formula InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;/m1./s1
Formula Weight 471.2961
Percent Purity >95%
Chemical Name or Material Glycochenodeoxycholic Acid Sodium Salt
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.