missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A Antibody (27A8), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Glycophorin A |
|---|---|
| Clone | 27A8 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30023607
|
Novus Biologicals
NBP3-26243-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036572
|
Novus Biologicals
NBP3-26243-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Glycophorin A | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
| A synthesized peptide derived from Human Glycophorin A [UniProt P02724] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 27A8 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2993 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title