Learn More
GLYCTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12426-1-AP-150UL
 View more versions of this product
Product Code. 16800144
  • 410.00€ / 150µL
Description
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifications
Specifications
GLYCTK
Polyclonal
Unconjugated
GLYCTK
glycerate kinase, GLYCTK, HBeAg binding protein 4, HBeAgBP4A, HBEBP2, HBEBP4
Rabbit
Antigen Affinity Chromatography
RUO
132158, 235582, 684314
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q0VGK3, Q8IVS8, Q8QZY2
Glyctk
GLYCTK Fusion Protein Ag3097
150 μL
Primary
Human, Mouse, Rat