missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycine Benzyl Ester p-Toluenesulfonate, 97%, BLD Pharm™

Product Code. 30802022
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30802022 25 g 25g
30791365 100 g 100g
30786291 1 g 1g
30796956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30802022 Supplier BLD Pharm Supplier No. BD13229725g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1738-82-5
Molecular Formula C18H22N2O6S
MDL Number MFCD00050678
Quantity 25 g
SMILES O=C(OCC1=CC=CC=C1)CNC(CN)=O.O=S(C2=CC=C(C)C=C2)(O)=O
Molecular Weight (g/mol) 394.44
Percent Purity 97%
Chemical Name or Material Glycylglycine Benzyl Ester p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.