missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal Sodium Bisulfite (contains oligomers), TRC

Product Code. 30400697
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400697 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400697 Supplier Toronto Research Chemicals Supplier No. TRCG75451310G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyoxal Sodium Bisulfite
CAS 517-21-5
Molecular Formula C2H4Na2O8S2
Quantity 10 g
Synonym Glyoxal Sodium Bisulfite
InChI Formula InChI=1S/C2H6O8S2.2Na/c3-1(11(5,6)7)2(4)12(8,9)10;;/h1-4H,(H,5,6,7)(H,8,9,10);;/q;2*+1/p-2
SMILES [Na+].[Na+].OC(C(O)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name disodium;1,2-dihydroxyethane-1,2-disulfonate
Molecular Weight (g/mol) 266.15
Formula Weight 265.9143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.