missing translation for 'onlineSavingsMsg'
Learn More

Glyphosate, TRC

Product Code. 30004693
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004693 25 g 25g
18073530 250 mg 250mg
18020530 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004693 Supplier Toronto Research Chemicals Supplier No. TRCG76500025G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-83-6
Molecular Formula C3 H8 N O5 P
Molecular Weight (g/mol) 169.07
Synonym N-(Phosphonomethyl)glycine, (Carboxymethylamino)methylphosphonic acid, Accord 4L, Accord XRT II, Atila, Buccaneer, Buccaneer Plus, C-K Yuyos FAV, Carboxymethylaminomethanephosphinic acid, Folusen, Forsat, Gibson, Glialka, Glialka 36, Glifoglex, Glifosan 747, Gliz, Gliz 480CS, GlyGran, Glyfos, Glyphodin A, Glyphomax, Glyphosate, Glyphosate CT, Green Fire 50SL, Ground Bio, HM 2028, Herbatop, Hockey, Kickdown, Klinik, Lancer, Lancer (herbicide), Leone, MON 2139, MON 3539, MON 6000, N-Phosphomethylglycine, NSC 151063, Phorsat, Phosphonomethylglycine, Phosphonomethyliminoacetic acid, Rebel Garden, Roundup Max, Safal, Scout, Scout (herbicide), Silglif, Total, Weedmaster 50SL
IUPAC Name 2-(phosphonomethylamino)acetic acid
SMILES OC(=O)CNCP(=O)(O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)
Formula Weight 169.014
Percent Purity >95%
Chemical Name or Material Glyphosate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.