missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glypican 1 Monoclonal Antibody (1019718)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561451

Product Code. 30534945

  • 410.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glypican 1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P35052
GPC1
Mouse myeloma cell line NS0 dervived human Glypican 1, Asp24-Ser530.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry
1019718
Unconjugated
GPC1
AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1
Mouse
Protein A/G
RUO
2817
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.