missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 4 Antibody (51E3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
366.00€ - 463.00€
Specifications
| Antigen | Glypican 4 |
|---|---|
| Clone | 51E3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:200, ELISA, Immunocytochemistry/ Immunofluorescence 1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646136
|
Novus Biologicals
NBP2-49892-50ul |
50 μg |
366.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627536
|
Novus Biologicals
NBP2-49892-100ul |
100 μg |
463.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glypican 4 Monoclonal antibody specifically detects Glypican 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Glypican 4 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Stem Cells | |
| PBS (pH 7.4), 10% Glycerol | |
| 2239 | |
| IgG2a κ | |
| Protein A purified |
| 51E3 | |
| Western Blot 1:1000, Flow Cytometry 1:200, ELISA, Immunocytochemistry/ Immunofluorescence 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4) | |
| Recombinant human Glypican 4 (401-529aa) purified from E. coli | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title