Learn More
Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (1N3V8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558177
Description
Immunogen sequence: MNPSMKQKQE EIKENIKNSS VPRRTLKMIQ PSASGSLVGR ENELSAGLSK RKHRNDHLTS TTSSPGVIVP ESSENKNLGG VTQESFDLMI KENPSSQYWK EVAEKRRKAL YEALKENEKL HKEIEQKDNE IARLKKENKE LAEVAEHVQY MAELIERLNG EPLDNFESLD NQEFDSEEET VEDSLVEDSE IGTCAEGTVS SSTDAKPCI.
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.
Specifications
| GMNN | |
| Recombinant Monoclonal | |
| 0.63 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O75496, O88513 | |
| GMNN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-209 of human GMNN (NP_056979.1). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1N3V8 | |
| Unconjugated | |
| GMNN | |
| AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 291137, 51053, 57441 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.