missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ GMP Human AB Serum
Description
/////Used to formulate 2 liters of GMP Human T Cell Media at 5% supplementation/////cGMP, Xeno-free, and testing is compliant with requirements outlined in 21 CFR 610.40/////Extensive viral testing done at the donor and pool levels/////Small donor pool derived material///// Heat inactivation protocol available///// Have questions? See our FAQs
Specifications
Specifications
| Content And Storage | Store the unopened product frozen at ≤ -10°C. See Use by Date on the Certificate of Analysis. |
| Quantity | 100 mL |
| Source | Human |
| Product Line | R&D Systems |
| Regulatory Status | GMP |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?